Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610191 | ||||||||||||||||||||
Literature Links: |
AQP4-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP4-AS1 - AQP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHST9 - carbohydrate sulfotransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256316.1 | 6115 | Intron | NP_001243245.1 | |||
NM_031422.5 | 6115 | Intron | NP_113610.2 | |||
XM_005258362.3 | 6115 | UTR 3 | XP_005258419.1 | |||
XM_006722555.3 | 6115 | Intron | XP_006722618.1 | |||
XM_017026033.1 | 6115 | UTR 3 | XP_016881522.1 | |||
XM_017026034.1 | 6115 | UTR 3 | XP_016881523.1 |