Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300879 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
XG - Xg blood group | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141919.1 | Intron | NP_001135391.1 | ||||
NM_001141920.1 | Intron | NP_001135392.1 | ||||
NM_175569.2 | Intron | NP_780778.1 | ||||
XM_005274587.3 | Intron | XP_005274644.1 | ||||
XM_011545575.2 | Intron | XP_011543877.1 | ||||
XM_017029787.1 | Intron | XP_016885276.1 |