Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611987 MIM: 601426 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS25 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRPS25 - mitochondrial ribosomal protein S25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022497.4 | 2507 | Intron | NP_071942.1 |
NR2C2 - nuclear receptor subfamily 2 group C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291694.1 | 2507 | UTR 3 | NP_001278623.1 | |||
NM_003298.4 | 2507 | UTR 3 | NP_003289.2 | |||
XM_011534058.2 | 2507 | UTR 3 | XP_011532360.1 | |||
XM_011534059.2 | 2507 | UTR 3 | XP_011532361.1 | |||
XM_011534061.2 | 2507 | UTR 3 | XP_011532363.1 | |||
XM_011534063.2 | 2507 | UTR 3 | XP_011532365.1 | |||
XM_011534064.2 | 2507 | UTR 3 | XP_011532366.1 | |||
XM_011534065.2 | 2507 | UTR 3 | XP_011532367.1 | |||
XM_011534066.2 | 2507 | UTR 3 | XP_011532368.1 | |||
XM_017007118.1 | 2507 | UTR 3 | XP_016862607.1 | |||
XM_017007119.1 | 2507 | UTR 3 | XP_016862608.1 | |||
XM_017007120.1 | 2507 | UTR 3 | XP_016862609.1 |
Set Membership: |
HapMap |