Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608734 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC39A12 - solute carrier family 39 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145195.1 | 372 | Silent Mutation | GAC,GAT | D,D 33 | NP_001138667.1 | |
NM_001282733.1 | 372 | Silent Mutation | GAC,GAT | D,D 33 | NP_001269662.1 | |
NM_001282734.1 | 372 | Intron | NP_001269663.1 | |||
NM_152725.3 | 372 | Silent Mutation | GAC,GAT | D,D 33 | NP_689938.2 |
Set Membership: |
HapMap |