Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603332 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAH1 - dynein axonemal heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015512.4 | Intron | NP_056327.4 | ||||
XM_017006129.1 | Intron | XP_016861618.1 | ||||
XM_017006130.1 | Intron | XP_016861619.1 | ||||
XM_017006131.1 | Intron | XP_016861620.1 | ||||
XM_017006132.1 | Intron | XP_016861621.1 | ||||
XM_017006133.1 | Intron | XP_016861622.1 |
Set Membership: |
HapMap |