Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615956 MIM: 177060 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC151 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC151 - coiled-coil domain containing 151 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKCSH - protein kinase C substrate 80K-H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001329.2 | Intron | NP_001001329.1 | ||||
NM_001289102.1 | Intron | NP_001276031.1 | ||||
NM_001289103.1 | Intron | NP_001276032.1 | ||||
NM_001289104.1 | Intron | NP_001276033.1 | ||||
NM_002743.3 | Intron | NP_002734.2 | ||||
XM_011528130.1 | Intron | XP_011526432.1 | ||||
XM_011528131.1 | Intron | XP_011526433.1 | ||||
XM_011528132.1 | Intron | XP_011526434.1 | ||||
XM_017026977.1 | Intron | XP_016882466.1 |
Set Membership: |
HapMap Validated |