Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611502 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPK - centromere protein K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPWD1 - peptidylprolyl isomerase domain and WD repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278926.1 | Intron | NP_001265855.1 | ||||
NM_001278927.1 | Intron | NP_001265856.1 | ||||
NM_001278929.1 | Intron | NP_001265858.1 | ||||
NM_015342.3 | Intron | NP_056157.1 | ||||
XM_005248474.3 | Intron | XP_005248531.1 | ||||
XM_006714583.3 | Intron | XP_006714646.1 | ||||
XM_011543295.1 | Intron | XP_011541597.1 | ||||
XM_017009286.1 | Intron | XP_016864775.1 |
Set Membership: |
HapMap |