Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCAAGTATAGATTATTTTTCATT[G/T]TTCTTCCAACCTTGGGGTTGTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTC-338M12.4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CTC-338M12.4 - uncharacterized LOC101928649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM52 - tripartite motif containing 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032765.2 | 2528 | Intron | NP_116154.1 | |||
XM_005266000.3 | 2528 | UTR 3 | XP_005266057.1 | |||
XM_017009991.1 | 2528 | Intron | XP_016865480.1 | |||
XM_017009992.1 | 2528 | UTR 3 | XP_016865481.1 | |||
XM_017009993.1 | 2528 | UTR 3 | XP_016865482.1 | |||
XM_017009994.1 | 2528 | UTR 3 | XP_016865483.1 |
TRIM52-AS1 - TRIM52 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |