Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGTCACCACCGCCACCACCACC[C/T]TGAATTATTCAAGACCCTCCAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101929696 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929696 - uncharacterized LOC101929696 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSD2 - pleckstrin and Sec7 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032289.2 | 797 | Intron | NP_115665.1 | |||
XM_011537696.2 | 797 | Intron | XP_011535998.1 | |||
XM_017009975.1 | 797 | Intron | XP_016865464.1 | |||
XM_017009976.1 | 797 | UTR 5 | XP_016865465.1 | |||
XM_017009977.1 | 797 | UTR 5 | XP_016865466.1 |