Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607769 MIM: 611048 MIM: 602309 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PLEKHA4 - pleckstrin homology domain containing A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R15A - protein phosphatase 1 regulatory subunit 15A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014330.3 | Intron | NP_055145.3 |
TULP2 - tubby like protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |