Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613478 MIM: 191318 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC106 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC106 - coiled-coil domain containing 106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
U2AF2 - U2 small nuclear RNA auxiliary factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012478.1 | Intron | NP_001012496.1 | ||||
NM_007279.2 | Intron | NP_009210.1 | ||||
XM_006722994.2 | Intron | XP_006723057.1 | ||||
XM_011526410.1 | Intron | XP_011524712.1 |