Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610534 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCPS PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCPS - decapping enzyme, scavenger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014026.4 | 1095 | Silent Mutation | CAA,CAG | Q,Q 105 | NP_054745.1 | |
XM_011542778.2 | 1095 | Silent Mutation | CAA,CAG | Q,Q 112 | XP_011541080.1 | |
XM_011542780.2 | 1095 | Intron | XP_011541082.1 | |||
XM_017017612.1 | 1095 | UTR 5 | XP_016873101.1 |
HP11014 - uncharacterized HP11014 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |