Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151440 MIM: 164005 MIM: 611669 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LYL1 - LYL1, basic helix-loop-helix family member | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005583.4 | 444 | UTR 5 | NP_005574.2 | |||
XM_005259912.3 | 444 | UTR 5 | XP_005259969.1 | |||
XM_017026816.1 | 444 | UTR 5 | XP_016882305.1 | |||
XM_017026817.1 | 444 | UTR 5 | XP_016882306.1 |
NFIX - nuclear factor I X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT1 - tRNA methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |