Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609898 | ||||||||||||||||||||
Literature Links: |
C22orf31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C22orf31 - chromosome 22 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015370.1 | Intron | NP_056185.1 | ||||
XM_005261491.3 | Intron | XP_005261548.1 | ||||
XM_011530096.2 | Intron | XP_011528398.1 | ||||
XM_017028741.1 | Intron | XP_016884230.1 |
KREMEN1 - kringle containing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039570.2 | Intron | NP_001034659.2 | ||||
NM_032045.4 | Intron | NP_114434.3 | ||||
XM_011530429.2 | Intron | XP_011528731.1 | ||||
XM_011530430.2 | Intron | XP_011528732.1 | ||||
XM_011530431.2 | Intron | XP_011528733.1 | ||||
XM_011530432.2 | Intron | XP_011528734.1 | ||||
XM_017028989.1 | Intron | XP_016884478.1 |