Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602987 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC129 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC129 - coiled-coil domain containing 129 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257967.1 | Intron | NP_001244896.1 | ||||
NM_001257968.1 | Intron | NP_001244897.1 | ||||
NM_194300.3 | Intron | NP_919276.2 | ||||
XM_011515213.1 | Intron | XP_011513515.1 | ||||
XM_017011871.1 | Intron | XP_016867360.1 | ||||
XM_017011872.1 | Intron | XP_016867361.1 |
PDE1C - phosphodiesterase 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191056.3 | Intron | NP_001177985.1 | ||||
NM_001191057.3 | Intron | NP_001177986.1 | ||||
NM_001191058.3 | Intron | NP_001177987.2 | ||||
NM_001191059.3 | Intron | NP_001177988.1 | ||||
NM_001322055.1 | Intron | NP_001308984.1 | ||||
NM_001322056.1 | Intron | NP_001308985.1 | ||||
NM_001322057.1 | Intron | NP_001308986.1 | ||||
NM_001322058.1 | Intron | NP_001308987.1 | ||||
NM_001322059.1 | Intron | NP_001308988.1 | ||||
NM_005020.4 | Intron | NP_005011.1 | ||||
XM_017012264.1 | Intron | XP_016867753.1 | ||||
XM_017012265.1 | Intron | XP_016867754.1 | ||||
XM_017012266.1 | Intron | XP_016867755.1 | ||||
XM_017012267.1 | Intron | XP_016867756.1 |
Set Membership: |
HapMap |