Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608926 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EMID1 - EMI domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267895.1 | Intron | NP_001254824.1 | ||||
NM_133455.3 | Intron | NP_597712.2 | ||||
XM_005261328.2 | Intron | XP_005261385.1 | ||||
XM_005261329.2 | Intron | XP_005261386.1 | ||||
XM_011529868.2 | Intron | XP_011528170.1 | ||||
XM_011529869.2 | Intron | XP_011528171.1 | ||||
XM_011529870.2 | Intron | XP_011528172.1 | ||||
XM_011529871.2 | Intron | XP_011528173.1 | ||||
XM_011529872.2 | Intron | XP_011528174.1 | ||||
XM_011529873.2 | Intron | XP_011528175.1 | ||||
XM_011529874.2 | Intron | XP_011528176.1 | ||||
XM_011529875.2 | Intron | XP_011528177.1 | ||||
XM_011529876.1 | Intron | XP_011528178.1 | ||||
XM_017028589.1 | Intron | XP_016884078.1 |
Set Membership: |
HapMap Validated |