Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609584 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
L2HGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
L2HGDH - L-2-hydroxyglutarate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024884.2 | Intron | NP_079160.1 | ||||
XM_005268075.4 | Intron | XP_005268132.1 | ||||
XM_011537166.2 | Intron | XP_011535468.1 | ||||
XM_011537167.2 | Intron | XP_011535469.1 | ||||
XM_011537168.2 | Intron | XP_011535470.1 | ||||
XM_017021655.1 | Intron | XP_016877144.1 | ||||
XM_017021656.1 | Intron | XP_016877145.1 | ||||
XM_017021657.1 | Intron | XP_016877146.1 | ||||
XM_017021658.1 | Intron | XP_016877147.1 |
Set Membership: |
HapMap |