Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGAGTTTGACCAGATTGAAGT[A/G]CTCTTTCAGATCCTAGGAATCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194539 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF32 - zinc finger protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005368.2 | 2333 | Intron | NP_001005368.1 | |||
NM_001324164.1 | 2333 | Intron | NP_001311093.1 | |||
NM_001324165.1 | 2333 | Intron | NP_001311094.1 | |||
NM_001324166.1 | 2333 | Intron | NP_001311095.1 | |||
NM_001324167.1 | 2333 | Intron | NP_001311096.1 | |||
NM_001324168.1 | 2333 | Intron | NP_001311097.1 | |||
NM_006973.2 | 2333 | Intron | NP_008904.1 | |||
XM_017016616.1 | 2333 | UTR 5 | XP_016872105.1 |
ZNF32-AS1 - ZNF32 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF32-AS2 - ZNF32 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF32-AS3 - ZNF32 antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |