Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTGCTGAAATTAGATGGTATAA[C/T]ATTTGTCTAGTTGCTCAGTGATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP69 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP69 - ZFP69 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320178.1 | Intron | NP_001307107.1 | ||||
NM_001320179.1 | Intron | NP_001307108.1 | ||||
NM_198494.2 | Intron | NP_940896.2 | ||||
XM_005270809.4 | Intron | XP_005270866.1 | ||||
XM_006710605.3 | Intron | XP_006710668.1 | ||||
XM_006710606.3 | Intron | XP_006710669.1 |