Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606534 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A43 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP3A43 - cytochrome P450 family 3 subfamily A member 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278921.1 | Intron | NP_001265850.1 | ||||
NM_022820.4 | Intron | NP_073731.1 | ||||
NM_057095.2 | Intron | NP_476436.1 | ||||
NM_057096.3 | Intron | NP_476437.1 | ||||
XM_017012542.1 | Intron | XP_016868031.1 | ||||
XM_017012543.1 | Intron | XP_016868032.1 | ||||
XM_017012544.1 | Intron | XP_016868033.1 | ||||
XM_017012545.1 | Intron | XP_016868034.1 |
Set Membership: |
HapMap |