Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGCGCTGGAGCCAGGACCTGG[C/T]GTTGGGCGGTACTTGGACAGCGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194552 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF79 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF79 - zinc finger protein 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286696.1 | 48 | UTR 5 | NP_001273625.1 | |||
NM_001286697.1 | 48 | UTR 5 | NP_001273626.1 | |||
NM_001286698.1 | 48 | UTR 5 | NP_001273627.1 | |||
NM_001322260.1 | 48 | UTR 5 | NP_001309189.1 | |||
NM_007135.2 | 48 | UTR 5 | NP_009066.2 | |||
XM_006717279.3 | 48 | UTR 5 | XP_006717342.1 |
Set Membership: |
HapMap Validated |