Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGGAAAATGAAGAGGTAAAATA[C/T]ATTATGGCACTTATTGCTAGGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 166945 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929767 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929767 - uncharacterized LOC101929767 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NBR1 - NBR1, autophagy cargo receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291571.1 | Intron | NP_001278500.1 | ||||
NM_001291572.1 | Intron | NP_001278501.1 | ||||
NM_005899.4 | Intron | NP_005890.2 | ||||
NM_031862.3 | Intron | NP_114068.1 | ||||
XM_006721903.1 | Intron | XP_006721966.1 | ||||
XM_011524813.1 | Intron | XP_011523115.1 | ||||
XM_017024642.1 | Intron | XP_016880131.1 | ||||
XM_017024643.1 | Intron | XP_016880132.1 | ||||
XM_017024644.1 | Intron | XP_016880133.1 |