Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTGTTTAATTTTAGATGGCTT[C/T]GTGTATTCATTTTGGTTTCAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616945 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLVS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLVS2 - clavesin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010852.3 | 238 | Intron | NP_001010852.2 | |||
XM_017010266.1 | 238 | UTR 5 | XP_016865755.1 |
Set Membership: |
HapMap Validated |