Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610933 MIM: 180475 MIM: 194552 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRSAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRSAM1 - leucine rich repeat and sterile alpha motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005373.3 | Intron | NP_001005373.1 | ||||
NM_001005374.3 | Intron | NP_001005374.1 | ||||
NM_001190723.2 | Intron | NP_001177652.1 | ||||
NM_138361.5 | Intron | NP_612370.3 | ||||
XM_006717316.3 | Intron | XP_006717379.1 | ||||
XM_017015283.1 | Intron | XP_016870772.1 | ||||
XM_017015284.1 | Intron | XP_016870773.1 |
RPL12 - ribosomal protein L12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000976.3 | Intron | NP_000967.1 |
SNORA65 - small nucleolar RNA, H/ACA box 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF79 - zinc finger protein 79 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |