Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACCTCTGTGTTGGGGTTACCC[A/G]CAGGGAATCATGTCCATGAGATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600339 MIM: 611930 MIM: 611836 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDGF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HDGF - hepatoma-derived growth factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ISG20L2 - interferon stimulated exonuclease gene 20 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL24 - mitochondrial ribosomal protein L24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024540.3 | Intron | NP_078816.2 | ||||
NM_145729.2 | Intron | NP_663781.1 | ||||
XM_011509981.2 | Intron | XP_011508283.1 | ||||
XM_011509982.2 | Intron | XP_011508284.1 |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142560.1 | Intron | NP_001136032.1 | ||||
NM_015997.3 | Intron | NP_057081.3 | ||||
XM_005245234.4 | Intron | XP_005245291.1 | ||||
XM_005245235.4 | Intron | XP_005245292.1 | ||||
XM_005245236.4 | Intron | XP_005245293.1 | ||||
XM_005245237.4 | Intron | XP_005245294.1 | ||||
XM_011509600.1 | Intron | XP_011507902.1 | ||||
XM_017001404.1 | Intron | XP_016856893.1 | ||||
XM_017001405.1 | Intron | XP_016856894.1 | ||||
XM_017001406.1 | Intron | XP_016856895.1 |
Set Membership: |
HapMap Validated |