Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609709 MIM: 604641 MIM: 603360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf94 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf94 - chromosome 11 open reading frame 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LARGE2 - LARGE xylosyl- and glucuronyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK8IP1 - mitogen-activated protein kinase 8 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX16 - peroxisomal biogenesis factor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004813.2 | Intron | NP_004804.1 | ||||
NM_057174.2 | Intron | NP_476515.1 | ||||
XM_011520474.1 | Intron | XP_011518776.1 |