Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTATGCTTTGTCTTAGTCTTG[A/G]TCAGATGCAGGTATTCTCTGCATAA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 606079 | |||||||||||||||||||||||||||||
Literature Links: |
CD163L1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CD163L1 - CD163 molecule like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297650.1 | Intron | NP_001284579.1 | ||||
NM_174941.5 | Intron | NP_777601.2 | ||||
XM_011520616.1 | Intron | XP_011518918.1 | ||||
XM_011520617.2 | Intron | XP_011518919.1 | ||||
XM_011520618.2 | Intron | XP_011518920.1 | ||||
XM_011520619.1 | Intron | XP_011518921.1 | ||||
XM_011520620.1 | Intron | XP_011518922.1 | ||||
XM_011520621.2 | Intron | XP_011518923.1 |
Set Membership: |
HapMap |