Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608838 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC84214 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC84214 - uncharacterized LOC84214 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VKORC1L1 - vitamin K epoxide reductase complex subunit 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284342.1 | Intron | NP_001271271.1 | ||||
NM_173517.4 | Intron | NP_775788.2 | ||||
XM_011515830.2 | Intron | XP_011514132.1 | ||||
XM_011515831.2 | Intron | XP_011514133.1 |
Set Membership: |
HapMap |