Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTAGGGATCATTGTCTTGTTCCT[A/G]TATTTAATGGACATGTCTATATTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 305990 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLRA2 - glycine receptor alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001118885.1 | Intron | NP_001112357.1 | ||||
NM_001118886.1 | Intron | NP_001112358.1 | ||||
NM_001171942.1 | Intron | NP_001165413.1 | ||||
NM_002063.3 | Intron | NP_002054.1 | ||||
XM_006724487.3 | Intron | XP_006724550.1 | ||||
XM_011545495.2 | Intron | XP_011543797.1 | ||||
XM_011545496.2 | Intron | XP_011543798.1 | ||||
XM_017029427.1 | Intron | XP_016884916.1 | ||||
XM_017029428.1 | Intron | XP_016884917.1 | ||||
XM_017029429.1 | Intron | XP_016884918.1 |
Set Membership: |
HapMap |