Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGACACACAGTCATGGAAAGTT[C/T]TCAATGAATGTTAACTATTATTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609215 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100996654 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100996654 - uncharacterized LOC100996654 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC61G - Sec61 translocon gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012456.1 | Intron | NP_001012474.1 | ||||
NM_014302.3 | Intron | NP_055117.1 |
Set Membership: |
HapMap |