Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCAGTGGGATATCATCTCTCA[A/G]TGATTGGGAATTCTTCCTCGAGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300747 | ||||||||||||||||||||
Literature Links: |
STS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STS - steroid sulfatase (microsomal), isozyme S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000351.5 | Intron | NP_000342.2 | ||||
NM_001320750.1 | Intron | NP_001307679.1 | ||||
NM_001320751.1 | Intron | NP_001307680.1 | ||||
NM_001320752.1 | Intron | NP_001307681.1 | ||||
NM_001320753.1 | Intron | NP_001307682.1 | ||||
NM_001320754.1 | Intron | NP_001307683.1 |