Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGCAAGGCTCATTCCCGCCCA[A/G]CAAACACAGGTGTTGCTCTGGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605988 MIM: 614174 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLRE1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCLRE1C - DNA cross-link repair 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033855.2 | 149 | UTR 5 | NP_001029027.1 | |||
NM_001033857.2 | 149 | UTR 5 | NP_001029029.1 | |||
NM_001033858.2 | 149 | UTR 5 | NP_001029030.1 | |||
NM_001289076.1 | 149 | UTR 5 | NP_001276005.1 | |||
NM_001289077.1 | 149 | UTR 5 | NP_001276006.1 | |||
NM_001289078.1 | 149 | UTR 5 | NP_001276007.1 | |||
NM_001289079.1 | 149 | UTR 5 | NP_001276008.1 | |||
NM_022487.3 | 149 | UTR 5 | NP_071932.2 | |||
XM_006717491.3 | 149 | Intron | XP_006717554.1 | |||
XM_011519616.1 | 149 | Intron | XP_011517918.1 | |||
XM_011519617.1 | 149 | Intron | XP_011517919.1 | |||
XM_011519619.1 | 149 | Intron | XP_011517921.1 | |||
XM_011519620.2 | 149 | Intron | XP_011517922.1 | |||
XM_011519621.1 | 149 | Intron | XP_011517923.1 | |||
XM_017016556.1 | 149 | Intron | XP_016872045.1 | |||
XM_017016557.1 | 149 | Intron | XP_016872046.1 | |||
XM_017016558.1 | 149 | Intron | XP_016872047.1 | |||
XM_017016559.1 | 149 | Intron | XP_016872048.1 | |||
XM_017016560.1 | 149 | Intron | XP_016872049.1 |
MEIG1 - meiosis/spermiogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080836.2 | 149 | Intron | NP_001074305.1 |