Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCCTTTGCAACAAAAGGAGTG[A/G]TTTGCTAACTTTTTCTTGAGTAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607404 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFNLR1 - interferon lambda receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_170743.3 | 3183 | UTR 3 | NP_734464.1 | |||
NM_173064.2 | 3183 | UTR 3 | NP_775087.1 | |||
NM_173065.2 | 3183 | UTR 3 | NP_775088.1 | |||
XM_006710394.3 | 3183 | UTR 3 | XP_006710457.2 | |||
XM_017000479.1 | 3183 | UTR 3 | XP_016855968.1 |
Set Membership: |
HapMap |