Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604581 MIM: 615103 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFG3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AFG3L2 - AFG3 like matrix AAA peptidase subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006796.2 | 903 | UTR 3 | NP_006787.2 | |||
XM_011525601.2 | 903 | UTR 3 | XP_011523903.1 |
TUBB6 - tubulin beta 6 class V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303524.1 | 903 | Intron | NP_001290453.1 | |||
NM_001303525.1 | 903 | UTR 3 | NP_001290454.1 | |||
NM_001303526.1 | 903 | Intron | NP_001290455.1 | |||
NM_001303527.1 | 903 | Intron | NP_001290456.1 | |||
NM_001303528.1 | 903 | Intron | NP_001290457.1 | |||
NM_001303529.1 | 903 | Intron | NP_001290458.1 | |||
NM_001303530.1 | 903 | Intron | NP_001290459.1 | |||
NM_032525.2 | 903 | Intron | NP_115914.1 |
Set Membership: |
HapMap Validated |