Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATTTCATAATTTATATAGCAAT[C/T]TCAGAAGATTCACATATATCATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162641 MIM: 602001 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPY1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NPY1R - neuropeptide Y receptor Y1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000909.5 | Intron | NP_000900.1 | ||||
XM_005263031.3 | Intron | XP_005263088.1 | ||||
XM_011532010.2 | Intron | XP_011530312.1 |
NPY5R - neuropeptide Y receptor Y5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317091.1 | Intron | NP_001304020.1 | ||||
NM_001317092.1 | Intron | NP_001304021.1 | ||||
NM_006174.3 | Intron | NP_006165.1 | ||||
XM_005263038.3 | Intron | XP_005263095.1 | ||||
XM_011532015.2 | Intron | XP_011530317.1 | ||||
XM_011532017.2 | Intron | XP_011530319.1 | ||||
XM_017008255.1 | Intron | XP_016863744.1 | ||||
XM_017008256.1 | Intron | XP_016863745.1 |
Set Membership: |
HapMap Validated |