Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGATTTTCCTCTCATGCAGTAA[C/T]TTACACATTTTGTGTGGAGAAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603088 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FKBP15 - FK506 binding protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015258.1 | Intron | NP_056073.1 | ||||
XM_006717018.2 | Intron | XP_006717081.1 | ||||
XM_006717019.1 | Intron | XP_006717082.1 | ||||
XM_017014552.1 | Intron | XP_016870041.1 |
SLC31A2 - solute carrier family 31 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |