Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603402 MIM: 614309 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACOX3 - acyl-CoA oxidase 3, pristanoyl | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT44 - tRNA methyltransferase 44 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152544.2 | 744 | Intron | NP_689757.2 | |||
XM_005247950.3 | 744 | Intron | XP_005248007.1 | |||
XM_011513406.2 | 744 | Intron | XP_011511708.1 | |||
XM_011513407.2 | 744 | Intron | XP_011511709.1 | |||
XM_011513408.1 | 744 | UTR 5 | XP_011511710.1 | |||
XM_011513409.2 | 744 | UTR 5 | XP_011511711.1 | |||
XM_011513410.2 | 744 | Intron | XP_011511712.1 | |||
XM_017007811.1 | 744 | Intron | XP_016863300.1 |