Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMER2 - APC membrane recruitment protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152704.3 | 5235 | UTR 3 | NP_689917.2 | |||
NM_199138.2 | 5235 | UTR 3 | NP_954589.1 | |||
XM_005266279.3 | 5235 | UTR 3 | XP_005266336.1 | |||
XM_005266280.3 | 5235 | UTR 3 | XP_005266337.1 | |||
XM_017020415.1 | 5235 | UTR 3 | XP_016875904.1 |
LINC00463 - long intergenic non-protein coding RNA 463 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |