Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGAAGGGTCTTGTAAATGGAAG[C/T]ACAGAGGCTGAGTGCAATTGTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609383 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724404 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102724404 - uncharacterized LOC102724404 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIPAL4 - NIPA like domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099287.1 | Intron | NP_001092757.1 | ||||
NM_001172292.1 | Intron | NP_001165763.1 | ||||
XM_011534552.1 | Intron | XP_011532854.1 |
Set Membership: |
HapMap |