Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179512 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFHB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFHB - EF-hand domain family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144715.3 | Intron | NP_653316.3 | ||||
XM_005264889.1 | Intron | XP_005264946.1 | ||||
XM_011533382.1 | Intron | XP_011531684.1 | ||||
XM_011533383.2 | Intron | XP_011531685.2 | ||||
XM_011533385.2 | Intron | XP_011531687.1 | ||||
XM_017005741.1 | Intron | XP_016861230.1 | ||||
XM_017005742.1 | Intron | XP_016861231.1 | ||||
XM_017005743.1 | Intron | XP_016861232.1 | ||||
XM_017005744.1 | Intron | XP_016861233.1 | ||||
XM_017005745.1 | Intron | XP_016861234.1 |
RAB5A - RAB5A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292048.1 | Intron | NP_001278977.1 | ||||
NM_004162.4 | Intron | NP_004153.2 |
Set Membership: |
HapMap |