Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACTTTACCTGACAATGACGCCC[A/G]GCCGGAGGTCGAAGTTCTTATGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610550 | ||||||||||||||||||||
Literature Links: |
MAT1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAT1A - methionine adenosyltransferase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000429.2 | 1325 | Missense Mutation | CCG,CTG | P,L 357 | NP_000420.1 | |
XM_005269842.4 | 1325 | Missense Mutation | CCG,CTG | P,L 357 | XP_005269899.1 | |
XM_005269843.4 | 1325 | Missense Mutation | CCG,CTG | P,L 316 | XP_005269900.1 |