Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601470 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CX3CR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CX3CR1 - C-X3-C motif chemokine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171171.1 | 931 | Missense Mutation | ACG,ATG | T,M 280 | NP_001164642.1 | |
NM_001171172.1 | 931 | Missense Mutation | ACG,ATG | T,M 280 | NP_001164643.1 | |
NM_001171174.1 | 931 | Missense Mutation | ACG,ATG | T,M 312 | NP_001164645.1 | |
NM_001337.3 | 931 | Missense Mutation | ACG,ATG | T,M 280 | NP_001328.1 |