Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATAAAATGAATTATTGAATACCA[A/G]ATTATGTTTTAACAAGAACTGAAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hecw1 PubMed Links |
Hecw1 - HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081348.3 | Intron | NP_001074817.3 |