Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCGTGGTTATGGATCCTTTT[C/T]CCCAGGTTAGAATAATGCCATCTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adamdec1 PubMed Links |
Adamdec1 - ADAM-like, decysin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021475.2 | 920 | Silent Mutation | NP_067450.1 |