Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGGCGTATTAGCATGCCCATTA[A/G]TAGGAGAAAAAAAAAGAAAAAAAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ubap2 PubMed Links |
Ubap2 - ubiquitin-associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026872.1 | Intron | NP_081148.1 |