Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGTAAATGCAATTCTTTCTCC[A/G]CGCTCAGAAAGTGGAGGCCTTGGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pum2 PubMed Links |
Pum2 - pumilio RNA-binding family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160219.1 | 304 | Silent Mutation | NP_001153691.1 | |||
NM_001160220.1 | 304 | Silent Mutation | NP_001153692.1 | |||
NM_001160221.1 | 304 | Silent Mutation | NP_001153693.1 | |||
NM_001160222.1 | 304 | Silent Mutation | NP_001153694.1 | |||
NM_030723.2 | 304 | Silent Mutation | NP_109648.2 |