Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGGATGTGTGTGTTAGAGTAA[G/T]CCCTTCCTGCCTTTTGGAATGACAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Snap25 PubMed Links |
Snap25 - synaptosomal-associated protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011428.3 | Intron | NP_035558.1 |