Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATAGACAAGGAGATTATTTTAA[G/T]TACATACAGTCAAGTGCTGCAAAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Stk31 PubMed Links |
Stk31 - serine threonine kinase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029916.2 | 1782 | Missense Mutation | NP_084192.2 |