Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm3045 PubMed Links |
Gm3045 - predicted gene 3045 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Slc25a48 - solute carrier family 25, member 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177809.4 | Intron | NP_808477.2 |