Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAATTTATACTTTGGGTCTGTT[C/G]TGTATTATAACCTTATCAGGCAACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sfxn1 PubMed Links |
Sfxn1 - sideroflexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027324.5 | Intron | NP_081600.1 |